Learn More
Innovative Research Inc Ms tPA Act Two Chn Rcmb 100ug
Catalog No. 502744133
Encompass_Preferred
Mouse tPA Active Two Chain Recombinant 100ug - Mouse tPA Active Two Chain Recombinant from Innovative Research has been recombinantly produced in CHO cells and purified using affinity chromatography then activated from single-chain form with immobilized plasmin. This is a frozen liquid buffered in 0.4M HEPES; 0.1M NaCl; pH 7.4 and a purity of 95%. This product is useful as a positive control for ELISA and western blot and for a variety of in vitro applications including cell differentiation, cell signaling, clotting assays, fibrinolysis, plasminogen activation, protein-protein interactions, receptor binding, thromboelastography, and thrombolysis. 95% two chain tPA. More Details Source Insect Cell Culture Purification Affinity purified Purity 95% Storage Conditions -70 C
Catalog No. 50-274-4133 Supplier Innovative Research Inc Supplier No. IMSTPAATCRC100UG
Mouse tPA Active Two Chain Recombinant 100ug - Mouse tPA Active Two Chain Recombinant from Innovative Research has been recombinantly produced in CHO cells and purified using affinity chromatography then activated from single-chain form with immobilized plasmin. This is a frozen liquid buffered in 0.4M HEPES; 0.1M NaCl; pH 7.4 and a purity of 95%. This product is useful as a positive control for ELISA and western blot and for a variety of in vitro applications including cell differentiation, cell signaling, clotting assays, fibrinolysis, plasminogen activation, protein-protein interactions, receptor binding, thromboelastography, and thrombolysis. 95% two chain tPA. More Details Source Insect Cell Culture Purification Affinity purified Purity 95% Storage Conditions -70 C