Learn More
Innovative Research Inc Ms Ttl Comp Component 3 Kit
Catalog No. 502744176
Encompass_Preferred
Mouse Total Complement Component 3 (C3) ELISA Kit - Mouse Total Complement Component 3 (C3) ELISA Kit from Innovative Research is intended for the quantitative determination of total C3 antigen in mouse plasma, serum, and cell culture samples. Strip well format. Reagents for up to 96 tests. Mouse C3 will bind to the capture antibody coated on the microtiter plate. After appropriate washing steps, peroxidase labeled polyclonal anti-mouse C3 antibody binds to the captured protein. Excess antibody is washed away and TMB substrate is used for color development at 450nm. Please note, pooled normal plasma from rat or rabbit resulted in significant color development.
Catalog No. 50-274-4176 Supplier Innovative Research Inc Supplier No. IMSC3KTT
Mouse Total Complement Component 3 (C3) ELISA Kit - Mouse Total Complement Component 3 (C3) ELISA Kit from Innovative Research is intended for the quantitative determination of total C3 antigen in mouse plasma, serum, and cell culture samples. Strip well format. Reagents for up to 96 tests. Mouse C3 will bind to the capture antibody coated on the microtiter plate. After appropriate washing steps, peroxidase labeled polyclonal anti-mouse C3 antibody binds to the captured protein. Excess antibody is washed away and TMB substrate is used for color development at 450nm. Please note, pooled normal plasma from rat or rabbit resulted in significant color development.