Learn More
Innovative Research Inc Ms Ttl Vtrnctn Kit
Catalog No. 502744128
Encompass_Preferred
Mouse Total Vitronectin ELISA Kit - Mouse Total Vitronectin ELISA Kit from Innovative Research is for the quantitative determination of total vitronectin antigen in biological fluids. Strip well format. Reagents for up to 96 tests.Mouse vitronectin will bind to the affinity purified capture antibody coated on the microtiter plate. Free and ligand bound enzyme will react with the antibody on the plate. After appropriate washing steps, biotinylated anti-mouse vitronectin primary antibody binds to the captured protein. Excess antibody is washed away and bound polyclonal antibody is reacted with peroxidase conjugated streptavidin. Following an additional washing step, TMB substrate is used for color development at 450nm.
Catalog No. 50-274-4128 Supplier Innovative Research Inc Supplier No. IMSVTNKTT
Mouse Total Vitronectin ELISA Kit - Mouse Total Vitronectin ELISA Kit from Innovative Research is for the quantitative determination of total vitronectin antigen in biological fluids. Strip well format. Reagents for up to 96 tests.Mouse vitronectin will bind to the affinity purified capture antibody coated on the microtiter plate. Free and ligand bound enzyme will react with the antibody on the plate. After appropriate washing steps, biotinylated anti-mouse vitronectin primary antibody binds to the captured protein. Excess antibody is washed away and bound polyclonal antibody is reacted with peroxidase conjugated streptavidin. Following an additional washing step, TMB substrate is used for color development at 450nm.