Learn More
Innovative Research Inc Ms X Hmn PK moAbs 10G3 1mg
Catalog No. 502744198
Encompass_Preferred
Mouse Anti Human Prekallikrein Monoclonal Clone 10G3 1mg - Mouse Anti Human Prekallikrein Monoclonal Clone 10G3 from Innovative Research is a monoclonal antibody in a Frozen liquid format, buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6. This antibody has been purified using protein G affinity chromatography. This product can be used in analytic tools such as ELISA, Western blot and Affinity purification. This product is an excellent reagent for affinity purification of prekallikrein from human plasma. Clone 10G3-42, isotype IgG1 kappa.
Catalog No. 50-274-4198 Supplier Innovative Research Inc Supplier No. IMSAHUPK10G3C1MG
Mouse Anti Human Prekallikrein Monoclonal Clone 10G3 1mg - Mouse Anti Human Prekallikrein Monoclonal Clone 10G3 from Innovative Research is a monoclonal antibody in a Frozen liquid format, buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6. This antibody has been purified using protein G affinity chromatography. This product can be used in analytic tools such as ELISA, Western blot and Affinity purification. This product is an excellent reagent for affinity purification of prekallikrein from human plasma. Clone 10G3-42, isotype IgG1 kappa.