Learn More
Innovative Research Inc Ms X Hmn tPA Hvy Chn 2A153 1mg
Catalog No. 502744196
Encompass_Preferred
Mouse Anti Human tPA Heavy Chain Monoclonal Clone 2A153 1mg - Mouse Anti Human tPA Heavy Chain Monoclonal Clone 2A153 from Innovative Research is a monoclonal antibody in a Frozen liquid format, buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6. This antibody has been purified using protein A affinity chromatography. This product can be used in analytic tools such as ELISA kits and Western blot. This product produces excellent western blots with free and complexed forms of tPA. This epitope is localized to the heavy chain of human tPA. This is Isotype IgG.
Catalog No. 50-274-4196 Supplier Innovative Research Inc Supplier No. IMSAHUTPAHC2A153C1MG
Mouse Anti Human tPA Heavy Chain Monoclonal Clone 2A153 1mg - Mouse Anti Human tPA Heavy Chain Monoclonal Clone 2A153 from Innovative Research is a monoclonal antibody in a Frozen liquid format, buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6. This antibody has been purified using protein A affinity chromatography. This product can be used in analytic tools such as ELISA kits and Western blot. This product produces excellent western blots with free and complexed forms of tPA. This epitope is localized to the heavy chain of human tPA. This is Isotype IgG.