Learn More
Julabo USA Inc MAGIO MS-Z 115V Bridge mounted Heating Circulator, 1kW to 300 °C, programmable, Pt100 port, USB/RS232/RS485/Ethernet connections, optional Analog module. EA
SDP
Catalog No. NC1921780
Encompass_Preferred
MAGIO™ MS-Z Bridge mounted heating circulator with immersion depth of 150mm (5.9") and extendable bridge (33 to 68 cm; 13" to 26.7") for tanks to 100 liters with 1 kW (115V) heating power to +300 °C. Large, color touchscreen UI with built-in help functions. For internal and external temperature applications. Includes Intelligent Cascade Control, self-optimizing for optimum results with settings for band limit, limit setting, co-speed factor and control dynamics. Variable speed pressure / suction pump 13-31 L/min; 3.5-13.3 PSI. Built-in cooling coil enables counter cooling with tap water. Barbed fittings for 8 or 12mm ID tubing included (pump connections M16x1 male). Includes Pt100 connection for external monitoring/control, RS232/RS485, USB and Ethernet ports with optional slot for analog communication (requires module 9900100). Modbus TCP/IP incorporated. Stakei port for external accessory control (solenoid valve, etc). Class III (FL) according to DIN 12876-1.
Catalog No. NC1921780 Supplier Julabo USA Inc Supplier No. 9032201.22
Add to Cart
Add to Cart
Request a Quote
MAGIO™ MS-Z Bridge mounted heating circulator with immersion depth of 150mm (5.9") and extendable bridge (33 to 68 cm; 13" to 26.7") for tanks to 100 liters with 1 kW (115V) heating power to +300 °C. Large, color touchscreen UI with built-in help functions. For internal and external temperature applications. Includes Intelligent Cascade Control, self-optimizing for optimum results with settings for band limit, limit setting, co-speed factor and control dynamics. Variable speed pressure / suction pump 13-31 L/min; 3.5-13.3 PSI. Built-in cooling coil enables counter cooling with tap water. Barbed fittings for 8 or 12mm ID tubing included (pump connections M16x1 male). Includes Pt100 connection for external monitoring/control, RS232/RS485, USB and Ethernet ports with optional slot for analog communication (requires module 9900100). Modbus TCP/IP incorporated. Stakei port for external accessory control (solenoid valve, etc). Class III (FL) according to DIN 12876-1.