Learn More
TARGETMOL CHEMICALS INC MS0124 5MG
SDP
Catalog No. 502580549
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MS0124 is a potent and selective G9A-like protein (GLP) inhibitor with IC50 values of 13±4 nM and 440±63 nM respectively. purity: 99%
Catalog No. 50-258-0549 Supplier TARGETMOL CHEMICALS INC Supplier No. T281125MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MS0124 is a potent and selective G9A-like protein (GLP) inhibitor with IC50 values of 13±4 nM and 440±63 nM respectively. purity: 99%