Learn More
Medchemexpress LLC HY-19615 2mg , MS023 CAS:1831110-54-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY196152MG
Encompass_Preferred
HY-19615 2mg MS023 CAS: 1831110-54-3 MS023 is a potent, selective, and cell-active inhibitor of human type I protein arginine methyltransferases (PRMTs) inhibitor, with IC50s of 30, 119, 83, 4 and 5 nM for PRMT1, PRMT3, PRMT4, PRMT6, and PRMT8, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1045