Learn More
Apexbio Technology LLC MS023,5mg CAS# 1831110-54-3
SDP
Supplier:  Apexbio Technology LLC B61835
Encompass_Preferred
MS023 is a type I PRMTs inhibitor.Protein arginine methyltransferases (PRMTs) play a critical role in various biological processes. PRMT overexpression has been observed in a variety of human diseases including cancer. PRMTs has been divided into three categories: type I PRMTs for catalyzing mono- and asymmetric dimethylation of arginine residues, type II PRMTs for catalyzing mono- and symmetric dimethylation of arginine residues, and type III PRMT for catalyzing only monomethylation of arginine residues. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0077