Learn More
Medchemexpress LLC HY-19615B 10mg , MS023 (dihydrochloride) CAS:1992047-64-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY19615B10MG
Encompass_Preferred
HY-19615B 10mg MS023 (dihydrochloride) CAS: 1992047-64-9 MS023 dihydrochloride is a potent, selective, and cell-active inhibitor of human type I protein arginine methyltransferases (PRMTs) inhibitor, with IC50s of 30, 119, 83, 4 and 5 nM for PRMT1, PRMT3, PRMT4, PRMT6, and PRMT8, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1046