Learn More
Medchemexpress LLC HY-100360 5mg Medchemexpress, MS049 CAS:1502816-23-0 Purity:>98%
SDP
Catalog No. 501931787
Encompass_Preferred
Medchemexpress, HY-100360 5mg MS049 CAS:1502816-23-0 MS049 is a potent, selective, and cell-active dual inhibitor of PRMT4 and PRMT6 with IC 50 s of 34 nM and 43 nM, respectively. MS049 reduces levels of Med12me2a and H3R2me2a in HEK293 cells. MS049 is not toxic and do not affect the growth of HEK293 cells [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1787 Supplier Medchemexpress LLC Supplier No. HY1003605MG
Medchemexpress, HY-100360 5mg MS049 CAS:1502816-23-0 MS049 is a potent, selective, and cell-active dual inhibitor of PRMT4 and PRMT6 with IC 50 s of 34 nM and 43 nM, respectively. MS049 reduces levels of Med12me2a and H3R2me2a in HEK293 cells. MS049 is not toxic and do not affect the growth of HEK293 cells [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.