Learn More
Cayman Chemical MS049 (hydrochloride), Purity: ≥98%, CAS: 2095432-59-8, 10 mg
SDP
Catalog No. 502965038
Encompass_Preferred
A selective inhibitor of PRMT4 (IC50 = 34 nM) and PRMT6 (IC50 = 43 nM); reduces H3R2me2a in HEK293 cells (IC50 = 0.97 µM) and also, unexpectedly, to reduce H4R3me2a in HEK293 cells
Catalog No. 50-296-5038 Supplier Cayman Chemical Supplier No. 1834810 MG
A selective inhibitor of PRMT4 (IC50 = 34 nM) and PRMT6 (IC50 = 43 nM); reduces H3R2me2a in HEK293 cells (IC50 = 0.97 µM) and also, unexpectedly, to reduce H4R3me2a in HEK293 cells