Learn More
ATCC MS1 GNAQ Q209L; Pancreas; Mouse (Mus musculus)
Supplier:  ATCC CRL3530
Encompass
MS1 GNAQ Q209L is a mouse cell line that can be used as a model of vascular malformation in vivo, including Sturge-Weber disease, and can be used to find drugs that are synthetic lethal to mutant GNAQ.
Catalog No. 50-238-4914