Learn More
Alkali Scientific Alkali Scientific MS1000 SmartSeal™ Automated Plate Sealer, 115V
SDP
Catalog No. 503029649
Encompass_Preferred
The Alkali Scientific MS1000 SmartSeal™ Automated Plate Sealer provides fast, reproducible sealing of microplates. Unlike manual plate sealing, the SmartSeal ensures uniform sealing of all wells and consistency from one plate to the next.   Fast, reproducible sealing of well plates Adjustable time, temperature and pressure for optimized sealing User friendly controls, load plate and press run to seal Accepts a wide range of plate types and heights, including deep well storage plates, assay plates, pcr plates, etc.
Catalog No. 50-302-9649 Supplier Alkali Scientific Supplier No. RS1000
The Alkali Scientific MS1000 SmartSeal™ Automated Plate Sealer provides fast, reproducible sealing of microplates. Unlike manual plate sealing, the SmartSeal ensures uniform sealing of all wells and consistency from one plate to the next.   Fast, reproducible sealing of well plates Adjustable time, temperature and pressure for optimized sealing User friendly controls, load plate and press run to seal Accepts a wide range of plate types and heights, including deep well storage plates, assay plates, pcr plates, etc.