Learn More
Medchemexpress LLC MS15 TFA | C66H80F3N11O7S | 1 MG
SDP
Catalog No. 5000373070
Encompass_Preferred
MS15 TFA is a potent and selective PROTAC (proteolysis-targeting chimera) degrader that targets AKT. It effectively inhibits the activity of all three AKT isoforms (AKT1, AKT2, and AKT3) with varying potencies. This compound is suitable for research applications to study AKT's role in cellular processes.
  • It is a potent and selective AKT PROTAC degrader.
  • It inhibits AKT1 activity with an IC50 of 798 nM.
  • It inhibits AKT2 activity with an IC50 of 90 nM.
  • It inhibits AKT3 activity with an IC50 of 544 nM.
  • It is bioavailable in mice through intraperitoneal administration.
  • It is supplied as a solid with a white to yellow appearance.
  • It is soluble in DMSO at concentrations of 100 mg/mL or higher.
Catalog No. 50-003-73070 Supplier Medchemexpress LLC Supplier No. HY151613A1MG
MS15 TFA is a potent and selective PROTAC (proteolysis-targeting chimera) degrader that targets AKT. It effectively inhibits the activity of all three AKT isoforms (AKT1, AKT2, and AKT3) with varying potencies. This compound is suitable for research applications to study AKT's role in cellular processes.
  • It is a potent and selective AKT PROTAC degrader.
  • It inhibits AKT1 activity with an IC50 of 798 nM.
  • It inhibits AKT2 activity with an IC50 of 90 nM.
  • It inhibits AKT3 activity with an IC50 of 544 nM.
  • It is bioavailable in mice through intraperitoneal administration.
  • It is supplied as a solid with a white to yellow appearance.
  • It is soluble in DMSO at concentrations of 100 mg/mL or higher.