Learn More
TARGETMOL CHEMICALS INC MS1943 5MG
SDP
Catalog No. 502581330
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MS1943 is a orally bioavailable EZH2 selective degrader(IC50 of 120 nM). purity: 98%
Catalog No. 50-258-1330 Supplier TARGETMOL CHEMICALS INC Supplier No. T137805MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MS1943 is a orally bioavailable EZH2 selective degrader(IC50 of 120 nM). purity: 98%