Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix MS2 Bacteriophage Extraction Control
GSA_VASDP

Catalog No. 22157429 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-429 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-157-429 Supplier ZeptoMetrix Supplier No. 0810066
 Restricted Item
Add to Cart
Add to Cart

Can be used as an internal control in nucleic acid-based amplification assays

  • Each frozen aliquot contains 1mL of partially purified, live, titered MS2 Bacteriophage
  • Single stranded RNA virus that infects Escherichia coli
  • Biosafety level 1 microorganism
  • Should be used within the confines of a Biological Safety cabinet

Controls should be run using the same protocols as those used to amplify extracted clinical specimens.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Biosafety Level 1
Certifications/Compliance ISO, 13485
Concentration 1 x 109PFU/mL
Content And Storage -20°C
Form Solution
Product Type MS2 Bacteriophage Extraction Control
For Use With (Application) Internal control in Nucleic Acid based amplification assays
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.