Learn More
ZeptoMetrix MS2 Bacteriophage-Medium
SDP
Catalog No. 22158034 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
22-158-034 1 mL
1 options
Catalog No. 22-158-034 Supplier ZeptoMetrix Supplier No. 0810274
Add to Cart
Add to Cart
Used as internal control in nucleic acid-based amplification assays to monitor reverse-transcription and PCR inhibition
  • Each frozen aliquot contains 0.1 or 1 mL of partially purified, live, titered MS2 Bacteriophage
  • Contain intact organisms and should be run in a manner identical to that used for clinical specimen
Specifications
Product Type MS2 Bacteriophage-Medium
Quantity 1 mL