Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix MS2 Bacteriophage, Titered
SDP

Catalog No. 22156880 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-156-880 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-156-880 Supplier ZeptoMetrix Supplier No. 0810052
Add to Cart
Add to Cart

Single stranded RNA virus that infects Escherichia coli

  • Can be used as an internal control in nucleic acid-based amplification assays
  • Each aliquot contains 0.1mL of partially purified, live, titered MS2 Bacteriophage, may also contain some E. coli DNA
  • The titer adjusted to 1.0 x 109 (plaque forming units/mL (pfu/mL)) prior to aliquoting and storage
  • Product supplied in SM buffer

Controls should be run using the same protocols as those used to amplify extracted clinical specimens.

Nucleic acid-based amplification assays

Specifications

Biosafety Level 1
Certifications/Compliance ISO, 13485
Concentration 1.0 x 109pfu/mL
Content And Storage -20°C
Form Solution
Product Type Titered Live Bacteria Control
For Use With (Application) Internal control in Nucleic Acid based amplification assays
Quantity 0.1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.