Learn More
Medchemexpress LLC HY-112620 10mg Medchemexpress, MS21570 CAS:65373-29-7 Purity:>98%
SDP
Catalog No. 501969120
Encompass_Preferred
Medchemexpress, HY-112620 10mg MS21570 CAS:65373-29-7 MS21570 is a selective GPR171 antagonist, with an IC50 of 220 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9120 Supplier Medchemexpress LLC Supplier No. HY11262010MG
Medchemexpress, HY-112620 10mg MS21570 CAS:65373-29-7 MS21570 is a selective GPR171 antagonist, with an IC50 of 220 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.