Learn More
Cayman Chemical MS-275, 209783-80-2, 5 mg
SDP
Supplier:  Cayman Chemical 132845
Encompass_Preferred
Molecular Formula C21H20N4O3, Purity ≥98%, Formula Weight 376.4, Synonyms: Entinostat, SNDX 275; An inhibitor of HDACs that preferentially inhibits HDAC1 (IC50 = 300 nM) over HDAC3 (IC50 = 8 M); does not inhibit HDAC8; induces p21/CIP1/WAF1, slowing cell growth, differentiation, and tumor development in vivo.
Catalog No. NC0922469
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.