Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MS402 | 1672684-68-2 | 98.4% | 10 MG
SDP

Catalog No. 5000234141
Encompass_Preferred

MS402 is a BD1-selective BET bromodomain inhibitor for research use, used to probe BET-dependent pathways in cellular and preclinical studies.

  • BD1-selective inhibitor with reported high affinity for BRD4 BD1 (reported Ki ≈ 77 nM).
  • Molecular formula C20H19ClN2O3; molecular weight 370.83 g·mol⁻¹.
  • Soluble in DMSO at 100 mg/mL; sonication may be required to fully dissolve.
  • Powder storage recommended at low temperatures; follow supplier guidance for shelf life.
  • Available in small research pack sizes suitable for assay preparation.

Catalog No. 50-002-34141 Supplier Medchemexpress LLC Supplier No. HY12000010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MS402 is a BD1-selective BET bromodomain inhibitor for research use, used to probe BET-dependent pathways in cellular and preclinical studies.

  • BD1-selective inhibitor with reported high affinity for BRD4 BD1 (reported Ki ≈ 77 nM).
  • Molecular formula C20H19ClN2O3; molecular weight 370.83 g·mol⁻¹.
  • Soluble in DMSO at 100 mg/mL; sonication may be required to fully dissolve.
  • Powder storage recommended at low temperatures; follow supplier guidance for shelf life.
  • Available in small research pack sizes suitable for assay preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.