Learn More
Medchemexpress LLC Ms402 | 1672684-68-2 | 98.4% | 50 MG
SDP
Catalog No. 5000234144
Encompass_Preferred
MS402 is a BD1-selective inhibitor of BET family bromodomains used as a research reagent to study epigenetic regulation, Th17 cell differentiation, and inflammatory disease models. It has molecular formula C20H19ClN2O3, molecular weight 370.83, CAS 1672684-68-2, and is supplied at high reported purity for laboratory research.
  • Selective BD1 BET bromodomain inhibition with low- to mid-nanomolar Ki values.
  • Blocks Th17 cell differentiation and shows efficacy in colitis mouse models.
  • White to off-white solid; recommended storage: powder at -20°C, in solvent at -80°C.
  • Reported purity approximately 98.36%.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-34144 Supplier Medchemexpress LLC Supplier No. HY12000050MG
MS402 is a BD1-selective inhibitor of BET family bromodomains used as a research reagent to study epigenetic regulation, Th17 cell differentiation, and inflammatory disease models. It has molecular formula C20H19ClN2O3, molecular weight 370.83, CAS 1672684-68-2, and is supplied at high reported purity for laboratory research.
  • Selective BD1 BET bromodomain inhibition with low- to mid-nanomolar Ki values.
  • Blocks Th17 cell differentiation and shows efficacy in colitis mouse models.
  • White to off-white solid; recommended storage: powder at -20°C, in solvent at -80°C.
  • Reported purity approximately 98.36%.
  • For research use only; not for human or veterinary use.