Learn More
Medchemexpress LLC HY-120000 5mg Medchemexpress, MS402 CAS:1672684-68-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1200005MG
Encompass_Preferred
Medchemexpress, HY-120000 5mg MS402 CAS:1672684-68-2 MS402 is a BD1-selective BET BrD inhibitor with Kis of 77 nM, 718 nM, 110 nM, 200 nM, 83 nM, and 240 nM for BRD4(BD1), BRD4(BD2), BRD3(BD1), BRD3(BD2), BRD2(BD1) and BRD2(BD2), respectively. MS402 blocks Th17 cell differentiation and ameliorates colitis in mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0933