Learn More
Medchemexpress LLC HY-112155 5mg Medchemexpress, MS4078 CAS:2229036-62-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1121555MG
Encompass_Preferred
Medchemexpress, HY-112155 5mg MS4078 CAS:2229036-62-6 MS4078 is an anaplastic lymphoma kinase (ALK) PROTAC (degrader) with a Kd of 19 nM for binding affinity to ALK. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9042