Learn More
Medchemexpress LLC HY-111139 10mg Medchemexpress, MS417 CAS:916489-36-6 Purity:>98%
SDP
Catalog No. 501968827
Encompass_Preferred
Medchemexpress, HY-111139 10mg MS417 CAS:916489-36-6 MS417 is a BET-specific BRD4 inhibitor, binds to BRD4-BD1 and BRD4-BD2 with IC50s of 30, 46 nM and Kds of 36.1, 25.4 nM, respectively, with weak selectivity at CBP BRD (IC50, 32.7 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8827 Supplier Medchemexpress LLC Supplier No. HY11113910MG
Medchemexpress, HY-111139 10mg MS417 CAS:916489-36-6 MS417 is a BET-specific BRD4 inhibitor, binds to BRD4-BD1 and BRD4-BD2 with IC50s of 30, 46 nM and Kds of 36.1, 25.4 nM, respectively, with weak selectivity at CBP BRD (IC50, 32.7 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.