Learn More
Medchemexpress LLC MS4322 (isomer) | 2601727-80-2 | 99.2% | 1101.32 g/mol | C55H76N10O12S | 5 MG
SDP
Catalog No. 5000221360
Encompass_Preferred
MS4322 (isomer) is an isomeric PROTAC degrader that targets PRMT5, promoting its ubiquitination and proteasomal degradation for use in cellular and biochemical studies of PRMT5-related pathways.
  • Promotes ubiquitination and degradation of PRMT5 in cells.
  • Reduces PRMT5 protein with a DC50 of 1.1 μM in MCF-7 cells.
  • Inhibits PRMT5 methyltransferase activity with an IC50 of 18 nM.
  • High purity (99.22%) suitable for biochemical assays.
  • Available in small research quantities, including 5 MG packs.
  • For research use only; not for human therapeutic use.
Catalog No. 50-002-21360 Supplier Medchemexpress LLC Supplier No. HY141877B5MG
MS4322 (isomer) is an isomeric PROTAC degrader that targets PRMT5, promoting its ubiquitination and proteasomal degradation for use in cellular and biochemical studies of PRMT5-related pathways.
  • Promotes ubiquitination and degradation of PRMT5 in cells.
  • Reduces PRMT5 protein with a DC50 of 1.1 μM in MCF-7 cells.
  • Inhibits PRMT5 methyltransferase activity with an IC50 of 18 nM.
  • High purity (99.22%) suitable for biochemical assays.
  • Available in small research quantities, including 5 MG packs.
  • For research use only; not for human therapeutic use.