Learn More
Medchemexpress LLC MS4322 (isomer) | 2601727-80-2 | 99.9% | C55H76N10O12S | 25 MG
SDP
Catalog No. 5000352623
Encompass_Preferred
MS4322 (isomer) is an isomer of MS4322. This compound is a specific PRMT5 PROTAC degrader that reduces PRMT5 protein levels and inhibits its methyltransferase activity. It promotes ubiquitination and degradation of PRMT5. This product can be used for research in breast cancer, lung cancer, and hepatocellular cancer.
  • Specific PRMT5 PROTAC degrader
  • Reduces PRMT5 protein level
  • Inhibits methyltransferase activity of PRMT5
  • Promotes ubiquitination and degradation of PRMT5
  • Used for research in breast cancer, lung cancer, and hepatocellular cancer
Catalog No. 50-003-52623 Supplier Medchemexpress LLC Supplier No. 25MGHY141877B
MS4322 (isomer) is an isomer of MS4322. This compound is a specific PRMT5 PROTAC degrader that reduces PRMT5 protein levels and inhibits its methyltransferase activity. It promotes ubiquitination and degradation of PRMT5. This product can be used for research in breast cancer, lung cancer, and hepatocellular cancer.
  • Specific PRMT5 PROTAC degrader
  • Reduces PRMT5 protein level
  • Inhibits methyltransferase activity of PRMT5
  • Promotes ubiquitination and degradation of PRMT5
  • Used for research in breast cancer, lung cancer, and hepatocellular cancer