Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MS438 10mg | 512840-45-8 | 10 MG
SDP

Catalog No. 5000307559
Encompass_Preferred

MS438 is a small-molecule thyrotropin receptor (TSHR) agonist with nanomolar potency (EC50 = 53 nM) that binds the serpentine transmembrane portion of TSHR and induces T4 secretion in mice. It is supplied as a white to off-white solid with high purity for research use.

  • Potent TSHR agonist with EC50 of 53 nM.
  • Binds the serpentine (transmembrane) portion of TSHR.
  • Induces T4 secretion in mouse models.
  • High purity (99.85%) white to off-white solid.
  • Soluble in DMSO at ≥100 mg/mL; compatible with 10% DMSO/90% corn oil for in vivo dosing.
  • Recommended storage: powder at -20°C long term; stock solutions at -80°C.

Catalog No. 50-003-07559 Supplier Medchemexpress LLC Supplier No. HY11491810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MS438 is a small-molecule thyrotropin receptor (TSHR) agonist with nanomolar potency (EC50 = 53 nM) that binds the serpentine transmembrane portion of TSHR and induces T4 secretion in mice. It is supplied as a white to off-white solid with high purity for research use.

  • Potent TSHR agonist with EC50 of 53 nM.
  • Binds the serpentine (transmembrane) portion of TSHR.
  • Induces T4 secretion in mouse models.
  • High purity (99.85%) white to off-white solid.
  • Soluble in DMSO at ≥100 mg/mL; compatible with 10% DMSO/90% corn oil for in vivo dosing.
  • Recommended storage: powder at -20°C long term; stock solutions at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.