Learn More
Aviva Systems Biology Corp MS4A3 Antibody - N-terminal region (ARP62012_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62012_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MS4A3. It was validated on Western Blot. . Alternate names: CD20L, HTM4ApplicationWB. Species reactivity: Human
Catalog No. 50-287-544