Learn More
Medchemexpress LLC HY-125232 5mg Medchemexpress, MS645 CAS:2250091-96-2 Purity:>98%
SDP
Catalog No. 501969688
Encompass_Preferred
Medchemexpress, HY-125232 5mg MS645 CAS:2250091-96-2 MS645 is a bivalent BET bromodomains (BrD) inhibitor with a Ki of 18.4 nM for BRD4-BD1/BD2. MS645 spatially constrains bivalent inhibition of BRD4 BrDs resulting in a sustained repression of BRD4 transcriptional activity in solid-tumor cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9688 Supplier Medchemexpress LLC Supplier No. HY1252325MG
Medchemexpress, HY-125232 5mg MS645 CAS:2250091-96-2 MS645 is a bivalent BET bromodomains (BrD) inhibitor with a Ki of 18.4 nM for BRD4-BD1/BD2. MS645 spatially constrains bivalent inhibition of BRD4 BrDs resulting in a sustained repression of BRD4 transcriptional activity in solid-tumor cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.