Learn More
Medchemexpress LLC HY-119053 5mg Medchemexpress, MS7972 CAS:352553-42-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1190535MG
Encompass_Preferred
Medchemexpress, HY-119053 5mg MS7972 CAS:352553-42-5 MS7972 is a small molecule that blocks human p53 and CREB binding protein association. MS7972 can almost completely block this BRD interaction at 50 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0922