Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MS8511 (hydrochloride) | 3031788-28-7 | C28H42ClN5O3 | 1 MG
SDP

Catalog No. 5000375251
Encompass_Preferred

MS8511 hydrochloride is a chemical compound for research use, identified as 2-Propenamide, N-[6-methoxy-4-[[1-(1-methylethyl)-4-piperidinyl]amino]-7-[3-(1-pyrrolidinyl)propoxy]-2-quinolinyl]-, hydrochloride (1:1). It appears as an off-white to light yellow solid and has a purity of 98.07% by LCMS. This product is intended for laboratory chemicals and manufacture of substances.

  • For research use only
  • Off-white to light yellow solid appearance
  • Purity of 98.07%
  • Stable under recommended storage conditions
  • Recommended storage for powder: -20°C for 3 years
  • Recommended storage for in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-75251 Supplier Medchemexpress LLC Supplier No. HY150510A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MS8511 hydrochloride is a chemical compound for research use, identified as 2-Propenamide, N-[6-methoxy-4-[[1-(1-methylethyl)-4-piperidinyl]amino]-7-[3-(1-pyrrolidinyl)propoxy]-2-quinolinyl]-, hydrochloride (1:1). It appears as an off-white to light yellow solid and has a purity of 98.07% by LCMS. This product is intended for laboratory chemicals and manufacture of substances.

  • For research use only
  • Off-white to light yellow solid appearance
  • Purity of 98.07%
  • Stable under recommended storage conditions
  • Recommended storage for powder: -20°C for 3 years
  • Recommended storage for in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.