Learn More
Selleck Chemical LLC MS8709 E1974-25MG
SDP
Catalog No. 5000039501
Encompass_Preferred
MS8709 (EX-A11295) is a first-in-class G9a/GLP proteolysis targeting chimera (PROTAC) degrader that induces G9a/GLP degradation in a concentration time and ubiquitin-proteasome system (UPS)-dependent manner and does not alter the mRNA expression of G9a/GLP over other methyltransferases It displays superior cell growth inhibition in prostate leukemia and lung cancer cells over parent G9a/GLP inhibitors
Catalog No. 50-000-39501 Supplier Selleck Chemical LLC Supplier No. E197425MG
MS8709 (EX-A11295) is a first-in-class G9a/GLP proteolysis targeting chimera (PROTAC) degrader that induces G9a/GLP degradation in a concentration time and ubiquitin-proteasome system (UPS)-dependent manner and does not alter the mRNA expression of G9a/GLP over other methyltransferases It displays superior cell growth inhibition in prostate leukemia and lung cancer cells over parent G9a/GLP inhibitors