Learn More
Medchemexpress LLC MS9427 TFA | 98.4% | 1107.50 | 25 MG
SDP
Catalog No. 5000362677
Encompass_Preferred
MS9427 TFA is a potent PROTAC EGFR degrader that selectively degrades mutant EGFR through both the ubiquitin/proteasome system (UPS) and autophagy/lysosome pathways. It inhibits the proliferation of NSCLC cells and can be used for anticancer research. MS9427 TFA has antiproliferative activity against HCC-827 cells, potently inducing EGFRDel19 degradation and inhibiting EGFR phosphorylation.
  • Potent PROTAC EGFR degrader
  • Selectively degrades mutant EGFR
  • Functions through ubiquitin/proteasome system (UPS) and autophagy/lysosome pathways
  • Inhibits proliferation of NSCLC cells
  • Supports anticancer research
  • Displays antiproliferative activity against HCC-827 cells
  • Induces EGFRDel19 degradation
  • Inhibits EGFR phosphorylation
Catalog No. 50-003-62677 Supplier Medchemexpress LLC Supplier No. 25MGHY147941A
MS9427 TFA is a potent PROTAC EGFR degrader that selectively degrades mutant EGFR through both the ubiquitin/proteasome system (UPS) and autophagy/lysosome pathways. It inhibits the proliferation of NSCLC cells and can be used for anticancer research. MS9427 TFA has antiproliferative activity against HCC-827 cells, potently inducing EGFRDel19 degradation and inhibiting EGFR phosphorylation.
  • Potent PROTAC EGFR degrader
  • Selectively degrades mutant EGFR
  • Functions through ubiquitin/proteasome system (UPS) and autophagy/lysosome pathways
  • Inhibits proliferation of NSCLC cells
  • Supports anticancer research
  • Displays antiproliferative activity against HCC-827 cells
  • Induces EGFRDel19 degradation
  • Inhibits EGFR phosphorylation