Learn More
Medchemexpress LLC HY-136927 100mg , MSA-2 CAS:129425-81-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY136927100MG
Encompass_Preferred
HY-136927 100mg MSA-2 CAS: 129425-81-6 MSA-2, a potent and orally available non-nucleotide STING agonist, is bound to STING as a noncovalent dimer with nanomolar affinity. MSA-2 shows EC50s of 8.3 and 24 μM for human STING isoforms WT and HAQ, respectively. MSA-2 stimulates interferon-β secretion in tumors, induces tumor regression with durable antitumor immunity, and synergizes with anti-PD-1 in syngeneic mouse tumor models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0799