Learn More
Medchemexpress LLC MSA-2 | 129425-81-6 | 99.8% | 294.32 | 200 MG
SDP
Catalog No. 5000348703
Encompass_Preferred
MSA-2 is a potent and orally available non-nucleotide STING agonist that binds to STING as a noncovalent dimer with nanomolar affinity. It stimulates interferon-β secretion in tumors, induces tumor regression with durable antitumor immunity, and synergizes with anti-PD-1 in syngeneic mouse tumor models.
  • Potent and orally available non-nucleotide STING agonist
  • Binds to STING as a noncovalent dimer with nanomolar affinity
  • Stimulates interferon-β secretion in tumors
  • Induces tumor regression with durable antitumor immunity
  • Synergizes with anti-PD-1 in syngeneic mouse tumor models
  • Achieves comparable exposure in tumor and plasma
  • Exhibits dose-dependent antitumor activity
  • Induces elevations of IFN-β, IL-6, and TNF-α in tumors
Catalog No. 50-003-48703 Supplier Medchemexpress LLC Supplier No. HY136927200MG
MSA-2 is a potent and orally available non-nucleotide STING agonist that binds to STING as a noncovalent dimer with nanomolar affinity. It stimulates interferon-β secretion in tumors, induces tumor regression with durable antitumor immunity, and synergizes with anti-PD-1 in syngeneic mouse tumor models.
  • Potent and orally available non-nucleotide STING agonist
  • Binds to STING as a noncovalent dimer with nanomolar affinity
  • Stimulates interferon-β secretion in tumors
  • Induces tumor regression with durable antitumor immunity
  • Synergizes with anti-PD-1 in syngeneic mouse tumor models
  • Achieves comparable exposure in tumor and plasma
  • Exhibits dose-dependent antitumor activity
  • Induces elevations of IFN-β, IL-6, and TNF-α in tumors