Learn More
MSC MSA Air Pressure Regulator, 0-160, Max 125 PSI, 1 ea
Catalog No. 5000770295
Encompass
This air pressure regulator is designed for constant flow airline systems, providing precise control over air pressure within a specified range.
  • Range of 0-160
  • Maximum pressure of 125 PSI
  • NIOSH approved
Catalog No. 50-007-70295 Supplier MSC Supplier No. 07108160
This air pressure regulator is designed for constant flow airline systems, providing precise control over air pressure within a specified range.
  • Range of 0-160
  • Maximum pressure of 125 PSI
  • NIOSH approved