Learn More
MSC MSA Comfo Respirator Cartridge, L-Profile, P100, 10/Pack
Catalog No. 5000793994
Encompass
The MSA Comfo Respirator Cartridge is a P100, low-profile filter designed for respiratory protection in various environments.
  • NIOSH approved
  • Dimensions: 3.228 in (82.000 mm) length, 3.228 in (82.000 mm) width, 0.827 in (21.000 mm) height
  • Weight: 0.088 lb (0.040 kg)
  • Supplied in a pack of 10 cartridges
Catalog No. 50-007-93994 Supplier MSC Supplier No. 46982997
The MSA Comfo Respirator Cartridge is a P100, low-profile filter designed for respiratory protection in various environments.
  • NIOSH approved
  • Dimensions: 3.228 in (82.000 mm) length, 3.228 in (82.000 mm) width, 0.827 in (21.000 mm) height
  • Weight: 0.088 lb (0.040 kg)
  • Supplied in a pack of 10 cartridges