Learn More
MSC MSA Full-Facepiece Respirator, Medium, Silicone, 4-Point Suspension, 1 ea
Catalog No. 5000769418
Encompass
The Advantage® 3200 Full-Facepiece Respirator is an air-purifying respirator designed for comprehensive respiratory protection.
  • Suspension type: 4-point
  • Full facepiece design
  • Manufacturer series: 3000 series
  • Medium size
  • Bayonet filter connection and cartridge type
  • Constructed from silicone material
  • Approved to ANSI Z87.1 and NIOSH standards
  • Includes Advantage European-style harness
Catalog No. 50-007-69418 Supplier MSC Supplier No. 01062207
The Advantage® 3200 Full-Facepiece Respirator is an air-purifying respirator designed for comprehensive respiratory protection.
  • Suspension type: 4-point
  • Full facepiece design
  • Manufacturer series: 3000 series
  • Medium size
  • Bayonet filter connection and cartridge type
  • Constructed from silicone material
  • Approved to ANSI Z87.1 and NIOSH standards
  • Includes Advantage European-style harness