Learn More
MSC MSA Hard Hat, Class E, 4-Point Suspension, Blue, 1 ea
Catalog No. 5000677987
Encompass
This hard hat features a full brim, blue color, and a ratchet adjustment. Constructed from polyethylene, it is rated as ANSI Type I and Class E. The hard hat is not vented but is slotted, and it includes Wheelbrator graphics. It comes with a 4-point suspension system.
  • Full brim type
  • Blue color
  • Ratchet adjustment type
  • Polyethylene material
  • ANSI Type I
  • Class E rating
  • Not vented
  • Slotted
  • Wheelbrator graphics
  • Four-point suspension
Catalog No. 50-006-77987 Supplier MSC Supplier No. 32364689
This hard hat features a full brim, blue color, and a ratchet adjustment. Constructed from polyethylene, it is rated as ANSI Type I and Class E. The hard hat is not vented but is slotted, and it includes Wheelbrator graphics. It comes with a 4-point suspension system.
  • Full brim type
  • Blue color
  • Ratchet adjustment type
  • Polyethylene material
  • ANSI Type I
  • Class E rating
  • Not vented
  • Slotted
  • Wheelbrator graphics
  • Four-point suspension