Learn More
MSC MSA Hard Hat, V-Gard Slotted Cap, 4-Point Ratchet, 1 ea
Numéro de catalogue. 5000687737
Encompass
This V-Gard Slotted Cap hard hat is designed for impact resistance, featuring a Hi-Vis Orange plastic construction. It is a Type 1, Class E hard hat with a 4-point ratchet suspension system, providing a secure and adjustable fit.
  • V-Gard slotted cap style
  • High-visibility orange color
  • Ratchet adjustment type
  • Impact-resistant application
  • Plastic material
  • ANSI type I
  • Class E rating
  • Not vented
  • Slotted design
  • No hard hat graphics
  • Complies with ANSI/ISEA Z89.1-2014 and CSA Z94.1-2005 standards
  • V-Gard series
  • Four-point suspension
  • Plastic suspension material
Numéro de catalogue. 50-006-87737 Fournisseur MSC Code fournisseur 67700625
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
This V-Gard Slotted Cap hard hat is designed for impact resistance, featuring a Hi-Vis Orange plastic construction. It is a Type 1, Class E hard hat with a 4-point ratchet suspension system, providing a secure and adjustable fit.
  • V-Gard slotted cap style
  • High-visibility orange color
  • Ratchet adjustment type
  • Impact-resistant application
  • Plastic material
  • ANSI type I
  • Class E rating
  • Not vented
  • Slotted design
  • No hard hat graphics
  • Complies with ANSI/ISEA Z89.1-2014 and CSA Z94.1-2005 standards
  • V-Gard series
  • Four-point suspension
  • Plastic suspension material
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.