Learn More
MSC MSA Respirator Cartridges, Comfo, Mercury Vapor, P100, 6/Pk.
Catalog No. 5000802929
Encompass
These respirator cartridges are designed for use with both half and full facepieces, providing protection against mercury vapor. They are P100 rated, ensuring a high level of filtration efficiency.
  • Designed for half and full facepiece respirators
  • P100 filtration efficiency
  • Protects against mercury vapor
  • NIOSH approved
  • Packaged as 6 cartridges per package
Catalog No. 50-008-02929 Supplier MSC Supplier No. 88157508
These respirator cartridges are designed for use with both half and full facepieces, providing protection against mercury vapor. They are P100 rated, ensuring a high level of filtration efficiency.
  • Designed for half and full facepiece respirators
  • P100 filtration efficiency
  • Protects against mercury vapor
  • NIOSH approved
  • Packaged as 6 cartridges per package