Learn More
MSC MSA Safety Half-Mask Respirator, SoftFeel Silicone, Black, 1 ea
Catalog No. 5000766348
Encompass
The Comfo Classic Half-Mask Respirator features a SoftFeel® facepiece, enhancing comfort by increasing the softness of both Hycar rubber and silicone materials available. Its unique face seal design ensures an exceptional and tight fit. This unit also includes a color-coded sizing system for easy identification and sorting.
  • SoftFeel facepiece increases rubber and silicone softness
  • Face seal design provides tight, comfortable fit
  • Color-coded sizing system makes sorting and identification easy
Catalog No. 50-007-66348 Supplier MSC Supplier No. 97283154
The Comfo Classic Half-Mask Respirator features a SoftFeel® facepiece, enhancing comfort by increasing the softness of both Hycar rubber and silicone materials available. Its unique face seal design ensures an exceptional and tight fit. This unit also includes a color-coded sizing system for easy identification and sorting.
  • SoftFeel facepiece increases rubber and silicone softness
  • Face seal design provides tight, comfortable fit
  • Color-coded sizing system makes sorting and identification easy