Learn More
MSC MSA Safety Staz-On Pin-Lock Suspension, V-Gard Standard, 1 ea
Catalog No. 5000794639
Encompass
The MSA Safety Staz-On Suspension is designed for V-Gard caps and hats, featuring a 4-point pin-lock system. It ensures a firm, comfortable, and secure fit, providing essential crown clearance for impact energy absorption.
  • Easy to operate pin-lock suspension
  • Includes 4-point webbed suspension with perforated vinyl sweatband
  • Nylon and plastic construction
  • Approved to ANSI/ISEA Z89.1-2014 standards
Catalog No. 50-007-94639 Supplier MSC Supplier No. 46856019
The MSA Safety Staz-On Suspension is designed for V-Gard caps and hats, featuring a 4-point pin-lock system. It ensures a firm, comfortable, and secure fit, providing essential crown clearance for impact energy absorption.
  • Easy to operate pin-lock suspension
  • Includes 4-point webbed suspension with perforated vinyl sweatband
  • Nylon and plastic construction
  • Approved to ANSI/ISEA Z89.1-2014 standards