Learn More
MSC MSA Sun Shield, for V-Gard Hats, Gray, 1 ea
Catalog No. 5000795684
Encompass
The sun shield is designed for individuals working outdoors in sunny conditions. It extends the brim of protective caps all around to protect the wearer's ears and neck from the sun's rays. The front portion features a smoke-tinted Lexan construction, which helps reduce the risk of skin cancer. The extended brim also aids in keeping wearers cooler and more comfortable throughout the day.
  • Easily attaches to helmets and caps
  • Polyurethane brim extender protects against sun and heat
  • Smoke-tinted Lexan front for extra protection
  • One size fits all design
  • Meets ANSI/ISEA Z89.1-2014 standards
  • Constructed from ABS and polyurethane materials
Catalog No. 50-007-95684 Supplier MSC Supplier No. 41739145
The sun shield is designed for individuals working outdoors in sunny conditions. It extends the brim of protective caps all around to protect the wearer's ears and neck from the sun's rays. The front portion features a smoke-tinted Lexan construction, which helps reduce the risk of skin cancer. The extended brim also aids in keeping wearers cooler and more comfortable throughout the day.
  • Easily attaches to helmets and caps
  • Polyurethane brim extender protects against sun and heat
  • Smoke-tinted Lexan front for extra protection
  • One size fits all design
  • Meets ANSI/ISEA Z89.1-2014 standards
  • Constructed from ABS and polyurethane materials