Learn More
MSC MSA V-Gard 500 Hard Hat, Vented, Class C, 1 ea
Catalog No. 5000769160
Encompass
The MSA V-Gard 500 Series hard hat is an impact-resistant, vented, Type 1, Class C hard hat featuring a 4-point plastic suspension system with ratchet adjustment. It is designed to fit head sizes from 6-1/2 to 8.
  • Impact resistant
  • Vented design
  • Type 1, Class C rated
  • Four-point plastic suspension
  • Ratchet adjustment
  • Cap brim style
  • Complies with ANSI/ISEA Z89.1-2014 and CSA Z94.1-2015 standards
Catalog No. 50-007-69160 Supplier MSC Supplier No. 80197882
The MSA V-Gard 500 Series hard hat is an impact-resistant, vented, Type 1, Class C hard hat featuring a 4-point plastic suspension system with ratchet adjustment. It is designed to fit head sizes from 6-1/2 to 8.
  • Impact resistant
  • Vented design
  • Type 1, Class C rated
  • Four-point plastic suspension
  • Ratchet adjustment
  • Cap brim style
  • Complies with ANSI/ISEA Z89.1-2014 and CSA Z94.1-2015 standards