Learn More
MSC MSA V-Gard H1 Safety Helmet, Novent, Fas-Trac III, 1 ea
Catalog No. 5000782602
Encompass
The V-Gard H1 Safety Helmet is designed for head protection, classified as a Type I, Class E hard hat. It features a Fas-Trac® III Pivot suspension and is not vented, with a shell service life of 5 years and suspension of 1 year.
  • Hard hat class E
  • Head size range 6 1/2-8
  • Four-point suspension system
  • Fas-Trac® III Pivot suspension type
  • Non-vented design
  • Polyethylene material
  • Shell service life 5 years, suspension 1 year
  • Weight 1.146 lb (0.520 kg)
  • Approved to ANSI/ISEA Z89.1-2014 and CAN/CSA Z94.1-2015 standards
Catalog No. 50-007-82602 Supplier MSC Supplier No. 19312024
The V-Gard H1 Safety Helmet is designed for head protection, classified as a Type I, Class E hard hat. It features a Fas-Trac® III Pivot suspension and is not vented, with a shell service life of 5 years and suspension of 1 year.
  • Hard hat class E
  • Head size range 6 1/2-8
  • Four-point suspension system
  • Fas-Trac® III Pivot suspension type
  • Non-vented design
  • Polyethylene material
  • Shell service life 5 years, suspension 1 year
  • Weight 1.146 lb (0.520 kg)
  • Approved to ANSI/ISEA Z89.1-2014 and CAN/CSA Z94.1-2015 standards