Learn More
MSC MSA V-Gard Hard Hat, Slotted Cap, Fas-Trac III, 1 ea
Catalog No. 5000789172
Encompass
The V-Gard Slotted Cap hard hat is designed for superior head protection in demanding environments. Constructed from high-density polyethylene with a UV inhibitor, it is impact-resistant and meets stringent safety standards for industrial use.
  • Type I, Class E protection
  • Impact-resistant design for superior top impact protection
  • Slotted cap style with a cap brim
  • Fas-Trac III ratchet suspension for comfort and secure fit
  • Made from virgin high-density polyethylene (HDPE) with UV inhibitor
  • Meets ANSI/ISEA Z89.1-2014 and CSA Z94.1-2005 standards
  • Fits head sizes 6-1/2 to 8
  • Four-point suspension
Catalog No. 50-007-89172 Supplier MSC Supplier No. 69763613
The V-Gard Slotted Cap hard hat is designed for superior head protection in demanding environments. Constructed from high-density polyethylene with a UV inhibitor, it is impact-resistant and meets stringent safety standards for industrial use.
  • Type I, Class E protection
  • Impact-resistant design for superior top impact protection
  • Slotted cap style with a cap brim
  • Fas-Trac III ratchet suspension for comfort and secure fit
  • Made from virgin high-density polyethylene (HDPE) with UV inhibitor
  • Meets ANSI/ISEA Z89.1-2014 and CSA Z94.1-2005 standards
  • Fits head sizes 6-1/2 to 8
  • Four-point suspension