Learn More
MSC MSA V-Gard Visor, Clear PC, 8x17x0.04 in, 1 ea
Catalog No. 5000787375
Encompass
The V-Gard Visor is a contoured, clear polycarbonate visor engineered for general purpose applications. It offers reliable face protection and is designed for compatibility with various headgear.
  • Contoured, clear polycarbonate construction
  • Designed for general purpose applications
  • Measures 8 x 17 x 0.04 in
  • One size fits all for universal compatibility
  • Meets ANSI/ISEA Z87.1-2020 standards for eye and face protection
  • Meets ANSI/ISEA Z89.1-2014 standards for head protection accessories
Catalog No. 50-007-87375 Supplier MSC Supplier No. 30825954
The V-Gard Visor is a contoured, clear polycarbonate visor engineered for general purpose applications. It offers reliable face protection and is designed for compatibility with various headgear.
  • Contoured, clear polycarbonate construction
  • Designed for general purpose applications
  • Measures 8 x 17 x 0.04 in
  • One size fits all for universal compatibility
  • Meets ANSI/ISEA Z87.1-2020 standards for eye and face protection
  • Meets ANSI/ISEA Z89.1-2014 standards for head protection accessories